Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265691.1 | NP_001252620.1 | 194 | 87 | ||
GenBank mRNA | JU325541.1 | - | 144 | 87 | ||
JV600316.1 | - | 194 | 87 | |||
JV669320.1 | - | 211 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |