Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015118605.2 | XP_014974091.1 | 732 | 89 | ||
XM_015118606.2 | XP_014974092.1 | 547 | 89 | |||
XM_015118607.2 | XP_014974093.1 | 1128 | 89 | |||
XM_015118608.2 | XP_014974094.1 | 994 | 89 | |||
XM_015118609.2 | XP_014974095.1 | 732 | 89 | |||
XM_015118610.2 | XP_014974096.1 | 547 | 89 | |||
GenBank mRNA | JU336277.1 | - | 453 | 89 | ||
JU477119.1 | - | 478 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Macaca mulatta |