Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM91A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260943.1 | NP_001247872.1 | 347 | 138 | ||
XM_015145940.2 | XP_015001426.2 | 440 | 138 | |||
XM_015145941.2 | XP_015001427.2 | 252 | 138 | |||
XM_028852237.1 | XP_028708070.1 | 701 | 138 | |||
XM_028852238.1 | XP_028708071.1 | 565 | 138 | |||
GenBank mRNA | JU333143.1 | - | 332 | 138 | ||
JU472538.1 | - | 347 | 138 | |||
JV643601.1 | - | 363 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |