Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLDN19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001087771.4 | XP_001087771.1 | 704 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |