Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: METTL15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260955.2 | NP_001247884.1 | 848 | 105 | ||
XM_028832545.1 | XP_028688378.1 | 995 | 105 | |||
XM_028832546.1 | XP_028688379.1 | 958 | 105 | |||
GenBank mRNA | JU473559.1 | - | 848 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |