Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HEPACAM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015116032.2 | XP_014971518.1 | 1382 | 140 | ||
GenBank mRNA | JU321167.1 | - | 1326 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |