Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP1R21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015112318.2 | XP_014967804.1 | 1173 | 113 | ||
XM_015112321.2 | XP_014967807.1 | 1209 | 113 | |||
XM_028831687.1 | XP_028687520.1 | 1022 | 113 | |||
XM_028831688.1 | XP_028687521.1 | 1156 | 113 | |||
XM_028831689.1 | XP_028687522.1 | 1018 | 113 | |||
XM_028831690.1 | XP_028687523.1 | 1195 | 113 | |||
XM_028831691.1 | XP_028687524.1 | 962 | 113 | |||
GenBank mRNA | JU332921.1 | - | 1105 | 113 | ||
JU474323.1 | - | 1063 | 113 | |||
JV706950.1 | - | 1127 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |