Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UNC5CL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015136305.2 | XP_014991791.1 | 1697 | 139 | ||
XM_028847598.1 | XP_028703431.1 | 1702 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |