Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAAH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001095907.4 | XP_001095907.1 | 869 | 113 | ||
XM_015127557.2 | XP_014983043.1 | 994 | 113 | |||
GenBank mRNA | JU481280.1 | - | 127 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |