Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001091413.4 | XP_001091413.2 | 1705 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |