Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUTM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001087474.4 | XP_001087474.4 | 1776 | 86 | ||
XM_015141800.2 | XP_014997286.2 | 1562 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Macaca mulatta |