Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BPIFA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243274.2 | NP_001230203.2 | 888 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |