Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTDH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266369.1 | NP_001253298.1 | 1851 | 73 | ||
XM_015145659.2 | XP_015001145.1 | 1971 | 73 | |||
XM_015145660.2 | XP_015001146.1 | 1782 | 73 | |||
XM_028852184.1 | XP_028708017.1 | 1905 | 73 | |||
XM_028852186.1 | XP_028708019.1 | 1815 | 73 | |||
XM_028852188.1 | XP_028708021.1 | 1716 | 73 | |||
GenBank mRNA | JU321225.1 | - | 1782 | 73 | ||
JU321226.1 | - | 1782 | 73 | |||
JU321229.1 | - | 1782 | 73 | |||
JU470512.1 | - | 1852 | 73 | |||
JV044132.1 | - | 1851 | 73 | |||
JV633395.1 | - | 1854 | 73 |