Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MICU3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028852366.1 | XP_028708199.1 | 736 | 134 | ||
XM_028852367.1 | XP_028708200.1 | 736 | 134 | |||
XM_028852369.1 | XP_028708202.1 | 736 | 134 | |||
GenBank mRNA | JU326223.1 | - | 469 | 134 | ||
JV614103.1 | - | 643 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |