Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC46A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015120809.2 | XP_014976295.2 | 1606 | 150 | ||
XM_028837533.1 | XP_028693366.1 | 2066 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |