Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HEATR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015144065.2 | XP_014999551.2 | 2709 | 79 | ||
XM_015144066.2 | XP_014999552.2 | 2744 | 79 | |||
XM_028851643.1 | XP_028707476.1 | 3073 | 79 | |||
XM_028851644.1 | XP_028707477.1 | 2938 | 79 | |||
XM_028851646.1 | XP_028707479.1 | 2743 | 79 | |||
XM_028851647.1 | XP_028707480.1 | 2746 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Macaca mulatta |