Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HEATR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015144065.2 | XP_014999551.2 | 2461 | 131 | ||
XM_015144066.2 | XP_014999552.2 | 2496 | 131 | |||
XM_028851643.1 | XP_028707476.1 | 2825 | 131 | |||
XM_028851644.1 | XP_028707477.1 | 2690 | 131 | |||
XM_028851645.1 | XP_028707478.1 | 2496 | 131 | |||
XM_028851646.1 | XP_028707479.1 | 2495 | 131 | |||
XM_028851647.1 | XP_028707480.1 | 2498 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |