Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001083788.4 | XP_001083788.3 | 1917 | 118 | ||
GenBank mRNA | JV648946.1 | - | 123 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |