Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DACT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001083736.4 | XP_001083736.2 | 518 | 60 | ||
GenBank mRNA | JV635990.1 | - | 342 | 60 | ||
JV708431.1 | - | 275 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |