Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR6X1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194286.1 | NP_001181215.1 | 898 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |