Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001098128.3 | XP_001098128.2 | 1167 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |