Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LPAR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001101441.3 | XP_001101441.1 | 2299 | 124 | ||
GenBank mRNA | JU327314.1 | - | 1484 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |