Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NHLH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001117555.4 | XP_001117555.1 | 271 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |