Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSKH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265966.1 | NP_001252895.1 | 1117 | 94 | ||
GenBank mRNA | JU335506.1 | - | 1114 | 94 | ||
JU473230.1 | - | 1114 | 94 | |||
JV047688.1 | - | 1117 | 94 | |||
JV656318.1 | - | 1121 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta |