Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR1Q1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193814.1 | NP_001180743.1 | 851 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |