Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFIT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001086665.4 | XP_001086665.1 | 953 | 70 | ||
GenBank mRNA | JU322557.1 | - | 103 | 70 | ||
JV577029.1 | - | 106 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Macaca mulatta |