Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCDHB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193851.3 | NP_001180780.3 | 213 | 164 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |