Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLCE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266557.1 | NP_001253486.1 | 1059 | 107 | ||
XM_028850199.1 | XP_028706032.1 | 591 | 107 | |||
GenBank mRNA | JU333908.1 | - | 1059 | 107 | ||
JU475095.1 | - | 1059 | 107 | |||
JV046705.1 | - | 917 | 107 | |||
JV647834.1 | - | 1062 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |