Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR10G7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194295.3 | NP_001181224.3 | 843 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |