Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAP1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198678.1 | NP_001185607.1 | 744 | 157 | ||
XM_015152172.2 | XP_015007658.1 | 826 | 157 | |||
XM_028829029.1 | XP_028684862.1 | 705 | 157 | |||
XM_028829030.1 | XP_028684863.1 | 691 | 157 | |||
XM_028829031.1 | XP_028684864.1 | 700 | 157 | |||
GenBank mRNA | CO648508.1 | - | 184 | 157 | ||
JU320583.1 | - | 723 | 157 | |||
JU320584.1 | - | 714 | 157 | |||
JU472436.1 | - | 798 | 157 | |||
JU472437.1 | - | 789 | 157 | |||
JV047820.1 | - | 790 | 157 | |||
JV047821.1 | - | 799 | 157 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |