Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015109893.1 | XP_014965379.1 | 206 | 146 | ||
GenBank mRNA | JU332514.1 | - | 172 | 146 | ||
JU473989.1 | - | 139 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |