Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTSG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001114339.4 | XP_001114339.2 | 105 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |