Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001097622.4 | XP_001097622.1 | 485 | 154 | ||
GenBank mRNA | JU335976.1 | - | 323 | 154 | ||
JU476857.1 | - | 325 | 154 | |||
JV068985.1 | - | 214 | 154 | |||
JV639114.1 | - | 336 | 154 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |