Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NANOG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001112791.4 | XP_001112791.1 | 157 | 178 | ||
XM_028830308.1 | XP_028686141.1 | 157 | 178 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |