Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266672.1 | NP_001253601.1 | 1002 | 120 | ||
GenBank mRNA | JU320890.1 | - | 778 | 120 | ||
JU471743.1 | - | 1024 | 120 | |||
JU471744.1 | - | 1024 | 120 | |||
JV044700.1 | - | 1002 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta Inventoried |