Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLR3H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266612.1 | NP_001253541.1 | 269 | 91 | ||
XM_015150399.2 | XP_015005885.1 | 252 | 91 | |||
XM_015150400.2 | XP_015005886.1 | 462 | 91 | |||
GenBank mRNA | JU333220.1 | - | 368 | 91 | ||
JU474539.1 | - | 372 | 91 | |||
JV046339.1 | - | 269 | 91 | |||
JV573333.1 | - | 359 | 91 | |||
JV639477.1 | - | 391 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |