Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC697687 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001085444.4 | XP_001085444.1 | 368 | 71 | ||
XM_028832947.1 | XP_028688780.1 | 303 | 71 | |||
GenBank mRNA | JV523673.1 | - | 232 | 71 | ||
JV699398.1 | - | 213 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta Inventoried |