Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDE9A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161449.1 | NP_001154921.1 | 269 | 65 | ||
XM_028844729.1 | XP_028700562.1 | 444 | 65 | |||
XM_028844730.1 | XP_028700563.1 | 177 | 65 | |||
GenBank mRNA | FJ914626.1 | - | 269 | 65 | ||
JU321678.1 | - | 271 | 65 | |||
JU475918.1 | - | 262 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |