Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR10Q1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193880.1 | NP_001180809.1 | 954 | 74 | ||
XM_015114099.2 | XP_014969585.1 | 2226 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta |