Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR4M1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193882.1 | NP_001180811.1 | 853 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Macaca mulatta |