Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR51E2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194559.1 | NP_001181488.1 | 202 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |