Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPYL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194874.2 | NP_001181803.2 | 3647 | 98 | ||
GenBank mRNA | JU340984.1 | - | 480 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta |