Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEBP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190951.2 | NP_001177880.2 | 261 | 73 | ||
XM_015144897.2 | XP_015000383.2 | 387 | 73 | |||
XM_015144898.2 | XP_015000384.2 | 407 | 73 | |||
GenBank mRNA | JV458402.1 | - | 233 | 73 | ||
JV643578.1 | - | 312 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |