Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204971.1 | NP_001191900.1 | 394 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |