Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OTX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177645.1 | NP_001171116.1 | 272 | 74 | ||
XM_015143755.2 | XP_014999241.1 | 567 | 74 | |||
XM_015143757.2 | XP_014999243.1 | 543 | 74 | |||
XM_028850243.1 | XP_028706076.1 | 464 | 74 | |||
XM_028850244.1 | XP_028706077.1 | 478 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta |