Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPRED1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204360.1 | NP_001191289.1 | 725 | 101 | ||
GenBank mRNA | HM370981.1 | - | 725 | 101 | ||
JU319878.1 | - | 518 | 101 | |||
JU319879.1 | - | 518 | 101 | |||
JU319880.1 | - | 518 | 101 | |||
JV046307.1 | - | 661 | 101 | |||
JV535066.1 | - | 755 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |