Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANAPC15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193608.1 | NP_001180537.1 | 450 | 94 | ||
XM_015115055.2 | XP_014970541.1 | 485 | 94 | |||
XM_028832849.1 | XP_028688682.1 | 472 | 94 | |||
GenBank mRNA | CO582475.1 | - | 454 | 94 | ||
JU333868.1 | - | 400 | 94 | |||
JU470775.1 | - | 500 | 94 | |||
JV046680.1 | - | 438 | 94 | |||
JV567090.1 | - | 406 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta |