Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194091.2 | NP_001181020.1 | 164 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Macaca mulatta |