Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FNTB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257705.1 | NP_001244634.1 | 235 | 93 | ||
XM_015143839.2 | XP_014999325.2 | 545 | 93 | |||
GenBank mRNA | JU320672.1 | - | 235 | 93 | ||
JU473133.1 | - | 260 | 93 | |||
JV056415.1 | - | 261 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |