Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGFR1OP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257773.1 | NP_001244702.1 | 550 | 89 | ||
GenBank mRNA | JU320035.1 | - | 550 | 89 | ||
JV045435.1 | - | 459 | 89 | |||
JV637318.1 | - | 671 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Macaca mulatta |